4-Nitro-α,α,α-trifluoro-o-toluic acid ≥97%

Synonyms: 4-Nitro-2-(trifluoromethyl)benzoic acid

638706-5G 638706-1G
220083-824EA 185.77 USD
220083-824 220083-823
4-Nitro-α,α,α-trifluoro-o-toluic acid ≥97%
4-Nitro-α,α,α-trifluoro-o-toluic acid
Formula: C₈H₄F₃NO₄
MW: 235.12 g/mol
MDL Number: MFCD04039246
CAS Number: 320-37-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR