3,5-Dimethyl-p-anisic acid ≥97%

Synonyms: 3,5-Dimethyl-p-anisic acid, 4-Methoxy-3,5-dimethylbenzoic acid, 3,5-Dimethyl-4-methoxybenzenecarboxylic acid, 3,5-Dimethyl-4-methoxybenzoic acid

638692-1G 638692-5G
220083-821EA 96.92 USD
220083-821 220083-822
3,5-Dimethyl-p-anisic acid ≥97%
3,5-Dimethyl-p-anisic acid
Formula: C₁₀H₁₂O₃
MW: 180.2 g/mol
Boiling Pt: 313 °C (1013 hPa)
Melting Pt: 196 - 198 °C
Density: 1.253 g/cm³ (20 °C)
MDL Number: MFCD00020309
CAS Number: 21553-46-8
EINECS: 244-441-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR