(±)-2-Phenylpiperazine

638641-5G 638641-25G
220083-813EA 152.92 USD
220083-813 220083-812
(±)-2-Phenylpiperazine
(±)-2-Phenylpiperazine
Formula: C₁₀H₁₄N₂
MW: 162.23 g/mol
MDL Number: MFCD01871362
CAS Number: 5271-26-1

Order Now

Specification Test Results

Assay 96%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR