4,5-Dibromo-o-xylene ≥97%

Warning

Synonyms: 1,2-Dibromo-4,5-dimethylbenzene, 4,5-Dibromo-1,2-dimethylbenzene

638447-1G 638447-5G
220083-789EA 58.58 USD
220083-789 220083-790
4,5-Dibromo-o-xylene ≥97%
4,5-Dibromo-o-xylene
Formula: C₈H₈Br₂
MW: 263.96 g/mol
MDL Number: MFCD00082743
CAS Number: 24932-48-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR