2-Chloro-4-(trifluoromethyl)benzenesulfonyl chloride ≥97%

Synonyms: 2-Chloro-4-(trifluoromethyl)benzene-1-sulphonyl chloride

638366-1G 638366-5G
220083-776EA 185.77 USD
220083-776 220083-777
2-Chloro-4-(trifluoromethyl)benzenesulfonyl chloride ≥97%
2-Chloro-4-(trifluoromethyl)benzenesulphonyl chloride
Formula: C₇H₃Cl₂F₃O₂S
MW: 279.07 g/mol
Boiling Pt: 70 - 71 °C (0.5 -rr)
MDL Number: MFCD00052912
CAS Number: 175205-54-6
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR