3-Cyanobenzenesulfonyl chloride ≥97%

Synonyms: 3-Cyanobenzene-1-sulphonyl chloride, 3-Cyanobenzene-1-sulphonylchloride

638358-1G 638358-5G
220083-774EA 185.77 USD
220083-774 220083-775
3-Cyanobenzenesulfonyl chloride ≥97%
3-Cyanobenzenesulphonyl chloride
Formula: C₇H₄ClNO₂S
MW: 201.63 g/mol
MDL Number: MFCD00203480
CAS Number: 56542-67-7
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR