4-Cyanobenzenesulfonyl chloride ≥97%

Synonyms: 4-Cyanobenzene-1-sulphonyl chloride

638331-1G 638331-5G
220083-772EA 185.77 USD
220083-772 220083-773
4-Cyanobenzenesulfonyl chloride ≥97%
4-Cyanobenzenesulphonyl chloride
Formula: C₇H₄ClNO₂S
MW: 201.63 g/mol
MDL Number: MFCD00052337
CAS Number: 49584-26-1
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR