4-Fluoro-o-toluic acid ≥95%

638323-1G 638323-5G
220083-770EA 66.12 USD
220083-770 220083-771
4-Fluoro-o-toluic acid ≥95%
4-Fluoro-o-toluic acid
Formula: C₈H₇FO₂
MW: 154.14 g/mol
MDL Number: MFCD04115880
CAS Number: 321-21-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR