5-Nitro-o-toluic acid ≥97%

Synonyms: 2-Methyl-5-nitrobenzoic acid

638307-1G 638307-5G
220083-767EA 185.77 USD
220083-767 220083-768
5-Nitro-o-toluic acid ≥97%
5-Nitro-o-toluic acid
Formula: C₈H₇NO₄
MW: 181.15 g/mol
Melting Pt: 179 - 181 °C
Density: 1.49 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007371
CAS Number: 1975-52-6
EINECS: 217-829-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR