2-Nitro-p-toluic acid ≥97%

638293-1G 638293-5G
220083-548EA 42.69 USD
220083-548 220083-549
2-Nitro-p-toluic acid ≥97%
2-Nitro-p-toluic acid
Formula: C₈H₇NO₄
MW: 181.15 g/mol
MDL Number: MFCD00033899
CAS Number: 27329-27-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR