2-(Dicyclohexylphosphino)biphenyl

Synonyms: (2-Biphenyl)dicyclohexylphosphine

638099-5G 638099-1G 638099-25G
220083-527EA 85.85 USD
220083-527 220083-525 220083-526
2-(Dicyclohexylphosphino)biphenyl
2-(Dicyclohexylphosphino)biphenyl
Formula: C₂₄H₃₁P
MW: 350.48 g/mol
MDL Number: MFCD01862441
CAS Number: 247940-06-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR