Potassium methyltrifluoroborate

Synonyms: Potassium trifluoro(methyl)borate, Methyltrifluoroboric acid potassium salt

637890-1G 637890-5G
220083-743EA 185.77 USD
220083-743 220083-744
Potassium methyltrifluoroborate
Potassium methyltrifluoroborate
Formula: CH₃BF₃K
MW: 121.94 g/mol
MDL Number: MFCD08276757
CAS Number: 13862-28-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR