4-Iodobiphenyl ≥97%

Synonyms: 4-Iodo-1,1'-biphenyl

637769-5G 637769-25G
220083-737EA 67.85 USD
220083-737 220083-736
4-Iodobiphenyl ≥97%
4-Iodobiphenyl
Formula: C₁₂H₉I
MW: 280.11 g/mol
Boiling Pt: 320 °C (1013 hPa)
Melting Pt: 112 °C
Density: 1.584 g/cm³ (20 °C)
MDL Number: MFCD00019028
CAS Number: 1591-31-7
EINECS: 216-469-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR