2,5-Dibromo-3-picoline

Synonyms: 2,5-Dibromo-3-methylpyridine

637661-1G 637661-5G
220083-728EA 30.15 USD
220083-728 220083-729
2,5-Dibromo-3-picoline
2,5-Dibromo-3-picoline
Formula: C₆H₅Br₂N
MW: 250.92 g/mol
MDL Number: MFCD02093085
CAS Number: 3430-18-0
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR