2-Chloro-5-pyridineboronic acid

Synonyms: 2-Chloropyridine-5-boronic acid, 2-Chloro-5-pyridinylboronic acid, 6-Chloro-3-pyridinylboronic acid, (6-Chloropyridin-3-yl)boronic acid, 6-Chloropyridine-3-boronic acid

637386-1G 637386-5G
220083-684EA 17.31 USD
220083-684 220083-685
2-Chloro-5-pyridineboronic acid
2-Chloro-5-pyridineboronic acid
Formula: C₅H₅BClNO₂
MW: 157.36 g/mol
MDL Number: MFCD03094998
CAS Number: 444120-91-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR