Cobalt(II,III) oxide

Warning

Synonyms: Tricobalt tetraoxide, Cobaltic tetraoxide

637025-100G 637025-250G 637025-25G
220083-665EA 380 USD
220083-665 220083-666 220083-667
Cobalt(II,III) oxide
Cobalt(II,III) oxide
Formula: Co₃O₄
MW: 240.8 g/mol
Melting Pt: 895 °C
Density: 6.005 g/cm³ (20 °C)
MDL Number: MFCD00010939
CAS Number: 1308-06-1
EINECS: 215-157-2
Merck Index: 13,02454

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR