2-Chloro-3-fluoropyridine

636576-5G 636576-25G
220083-609EA 67.63 USD
220083-609 220083-608
2-Chloro-3-fluoropyridine
2-Chloro-3-fluoropyridine
Formula: C₅H₃ClFN
MW: 131.54 g/mol
MDL Number: MFCD03095302
CAS Number: 17282-04-1
UN: 2810
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR