5-Bromo-2-fluorobenzoic acid ≥97%

Warning

636452-1G 636452-5G
220083-600EA 38.88 USD
220083-600 220083-601
5-Bromo-2-fluorobenzoic acid ≥97%
5-Bromo-2-fluorobenzoic acid
Formula: C₇H₄BrFO₂
MW: 219.01 g/mol
MDL Number: MFCD00143423
CAS Number: 146328-85-0
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR