5-Isoxazolecarboxylic acid

Synonyms: Isoxazole-5-carboxylic acid

636258-1G 636258-5G
220083-514EA 185.77 USD
220083-514 220083-515
5-Isoxazolecarboxylic acid
5-Isoxazolecarboxylic acid
Formula: C₄H₃NO₃
MW: 113.07 g/mol
MDL Number: MFCD00156151
CAS Number: 21169-71-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR