Isoxazole-5-carbonyl chloride

636231-1G 636231-5G
220083-512EA 88.31 USD
220083-512 220083-513
Isoxazole-5-carbonyl chloride
Isoxazole-5-carbonyl chloride
Formula: C₄H₂ClNO₂
MW: 131.52 g/mol
Boiling Pt: 74…76 °C (1013 hPa)
MDL Number: MFCD00067868
CAS Number: 62348-13-4
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR