5-Bromo-2-thiophenesulfonyl chloride

Synonyms: 5-Bromo-2-thiophenesulphonyl Chloride, 5-Bromothiophene-2-sulphonyl chloride

636223-5G 636223-25G
220083-511EA 150.77 USD
220083-511 220083-510
5-Bromo-2-thiophenesulfonyl chloride
5-Bromo-2-thiophenesulphonyl chloride
Formula: C₄H₂BrClO₂S₂
MW: 261.55 g/mol
MDL Number: MFCD00084909
CAS Number: 55854-46-1
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR