6-Chlorooxindole

Synonyms: 6-Chloro-2-oxindole

636215-25G 636215-5G
220083-507EA 44.15 USD
220083-507 220083-508
6-Chlorooxindole
6-Chlorooxindole
Formula: C₈H₆ClNO
MW: 167.59 g/mol
MDL Number: MFCD00209962
CAS Number: 56341-37-8
EINECS: 421-330-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR