Neodymium(III) oxide ≥99.9%

Synonyms: Dineodymium trioxide, Neodymium(III)-oxide, Neodymium oxide

634611-5G 634611-50G
220083-405EA 123.38 USD
220083-405 220083-404
Neodymium(III) oxide ≥99.9%
Neodymium(III) oxide
Formula: Nd₂O₃
MW: 336.48 g/mol
Boiling Pt: 3760 °C (1013 hPa)
Melting Pt: 2270 °C
Density: 7.24 g/cm³ (20 °C)
MDL Number: MFCD00011134
CAS Number: 1313-97-9
EINECS: 215-214-1
Merck Index: 14,06450

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR