5-Bromo-1,2,3-trimethoxybenzene ≥97%

Warning

Synonyms: 1-Bromo-3,4,5-trimethoxybenzene

634530-100G 634530-10G
220083-394EA 367.23 USD
220083-394 220083-395
5-Bromo-1,2,3-trimethoxybenzene ≥97%
5-Bromo-1,2,3-trimethoxybenzene
Formula: C₉H₁₁BrO₃
MW: 247.09 g/mol
MDL Number: MFCD00017169
CAS Number: 2675-79-8
EINECS: 220-223-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR