4-Pyridineboronic acid

Synonyms: 4-Pyridinylboronic acid, Pyridine-4-boronic acid, Pyridin-4-ylboronic acid

634492-1G 634492-25G 634492-5G
220083-389EA 28.66 USD
220083-389 220083-390 220083-391
4-Pyridineboronic acid
4-Pyridineboronic acid
Formula: C₅H₆BNO₂
MW: 122.92 g/mol (anhy)
MDL Number: MFCD01074545
CAS Number: 1692-15-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR