Calcium oxide ≥98%

Danger

Synonyms: Quicklime, Calcium oxide from marble, caustic, Lime

634182-25G 634182-100G
220083-359EA 289.23 USD
220083-359 220083-358
Calcium oxide ≥98%
Calcium oxide
Formula: CaO
MW: 56.08 g/mol
Boiling Pt: 2850 °C (1013 hPa)
Melting Pt: 2580 °C
Density: 3.38 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010911
CAS Number: 1305-78-8
EINECS: 215-138-9
Merck Index: 13,01691

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR