Disodium malonate ≥97.0%

63409-500G 63409-100G
220083-347EA 241.23 USD
220083-347 220083-346
Disodium malonate ≥97.0%
Disodium malonate
Formula: C₃H₂Na₂O₄
MDL Number: MFCD00002708
CAS Number: 141-95-7
EINECS: 205-514-0

Order Now

Specification Test Results

Assay 97.0% NT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR