4-Chloro-o-toluic acid ≥96%

Warning

634034-1G 634034-5G
220083-334EA 48.46 USD
220083-334 220083-335
4-Chloro-o-toluic acid ≥96%
4-Chloro-o-toluic acid
Formula: C₈H₇ClO₂
MW: 170.6 g/mol
MDL Number: MFCD00045852
CAS Number: 7499-07-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR