5-Methylisoxazole-4-carboxylic acid

633771-1G 633771-10G
220083-317EA 37.37 USD
220083-317 220083-316
5-Methylisoxazole-4-carboxylic acid
5-Methylisoxazole-4-carboxylic acid
Formula: C₅H₅NO₃
MW: 127.1 g/mol
MDL Number: MFCD00955655
CAS Number: 42831-50-5
EINECS: 222-289-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR