2,4,6-Trichloropyridine

633534-5G 633534-1G
220082-863EA 1195.38 USD
220082-863 220082-862
2,4,6-Trichloropyridine
2,4,6-Trichloropyridine
Formula: C₅H₂Cl₃N
MW: 182.44 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00971928
CAS Number: 16063-69-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR