2-Acetyl-4-methylthiophene

Synonyms: 1-(4-Methyl-2-thienyl)-1-ethanone, 1-(4-Methylthiophen-2-yl)ethanone

633046-5G 633046-25G
220083-236EA 95.52 USD
220083-236 220083-235
2-Acetyl-4-methylthiophene
2-Acetyl-4-methylthiophene
Formula: C₇H₈OS
MW: 140.21 g/mol
Boiling Pt: 108…109 °C (14 mmHg)
MDL Number: MFCD00027395
CAS Number: 13679-73-7
EINECS: 237-180-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR