3-Bromo-2-thenoic acid

Synonyms: 3-Bromo-2-thiophenecarboxylic acid, 3-Bromothiophene-2-carboxylic acid

633011-5G 633011-25G
220083-234EA 31.23 USD
220083-234 220083-233
3-Bromo-2-thenoic acid
3-Bromo-2-thenoic acid
Formula: C₅H₃BrO₂S
MW: 207.05 g/mol
MDL Number: MFCD00052291
CAS Number: 7311-64-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR