4-Bromo-2,5-difluorobenzenesulfonyl chloride ≥97%

632856-5G 632856-1G
220083-216EA 136.77 USD
220083-216 220083-215
4-Bromo-2,5-difluorobenzenesulfonyl chloride ≥97%
4-Bromo-2,5-difluorobenzenesulphonyl chloride
Formula: C₆H₂BrClF₂O₂S
MW: 291.5 g/mol
Boiling Pt: 76 °C (0.01 -rr)
Melting Pt: 34 - 36 °C
MDL Number: MFCD00042181
CAS Number: 207974-14-9
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR