2-Methoxynicotinic acid

Synonyms: 2-Methoxy-3-pyridinecarboxylic acid, 2-Methoxypyridine-3-carboxylic acid

632767-5G 632767-1G
220083-206EA 174.46 USD
220083-206 220083-205
2-Methoxynicotinic acid
2-Methoxynicotinic acid
Formula: C₇H₇NO₃
MW: 153.14 g/mol
MDL Number: MFCD00085110
CAS Number: 16498-81-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR