3-Nitrophenylboronic acid pinacol ester ≥97%

Synonyms: 4,4,5,5-Tetramethyl-2-(3-nitrophenyl)-1,3,2-dioxaborolane

632708-1G 632708-5G
220083-195EA 60.92 USD
220083-195 220083-196
3-Nitrophenylboronic acid pinacol ester ≥97%
3-Nitrophenylboronic acid pinacol ester
Formula: C₁₂H₁₆BNO₄
MW: 249.07 g/mol
MDL Number: MFCD05663879
CAS Number: 68716-48-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR