2,4-Difluorophenylboronic acid pinacol ester ≥97%

Synonyms: 2-(2,4-Difluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

632694-5G 632694-10G
220083-194EA 88.62 USD
220083-194 220083-193
2,4-Difluorophenylboronic acid pinacol ester ≥97%
2,4-Difluorophenylboronic acid pinacol ester
Formula: C₁₂H₁₅BF₂O₂
MW: 240.06 g/mol
MDL Number: MFCD05663880
CAS Number: 288101-48-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR