2-(2-Hydroxyphenyl)benzothiazole

Synonyms: 2-(1,3-Benzothiazol-2-yl)phenol

632589-25G 632589-5G
220083-172EA 87.88 USD
220083-172 220083-173
2-(2-Hydroxyphenyl)benzothiazole
2-(2-Hydroxyphenyl)benzothiazole
Formula: C₁₃H₉NOS
MW: 227.29 g/mol
MDL Number: MFCD00022869
CAS Number: 3411-95-8
EINECS: 222-299-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR