2-Bromonicotinic acid

Synonyms: 2-Bromo-3-pyridinecarboxylic acid, 2-Bromopyridine-3-carboxylic acid

632465-5G 632465-1G
220083-163EA 54.82 USD
220083-163 220083-162
2-Bromonicotinic acid
2-Bromonicotinic acid
Formula: C₆H₄BrNO₂
MW: 202.01 g/mol
MDL Number: MFCD01646068
CAS Number: 35905-85-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR