Indole-7-carbaldehyd

Synonyms: 1H-Indole-7-carbaldehyde, Indole-7-carboxaldehyde

632414-5G 632414-1G
220083-159EA 326.31 USD
220083-159 220083-158
Indole-7-carbaldehyd
Indole-7-carbaldehyd
Formula: C₉H₇NO
MW: 145.16 g/mol
MDL Number: MFCD01318152
CAS Number: 1074-88-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR