Indole-6-carbaldehyd

Synonyms: Indole-6-carboxaldehyde, 1H-Indole-6-carbaldehyde

632406-250MG 632406-1G
220083-157EA 185.77 USD
220083-157 220083-156
Indole-6-carbaldehyd
Indole-6-carbaldehyd
Formula: C₉H₇NO
MW: 145.16 g/mol
MDL Number: MFCD02179596
CAS Number: 1196-70-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR