Indium (III) oxide ≥99.9%

Synonyms: Indium trioxide, Diindium trioxide

632317-5G 632317-25G
220083-153EA 178.46 USD
220083-153 220083-152
Indium (III) oxide ≥99.9%
Indium (III) oxide
Formula: In₂O₃
MW: 277.64 g/mol
Melting Pt: 715.05 °C
Density: 7.18 g/cm³ (20 °C)
MDL Number: MFCD00011060
CAS Number: 1312-43-2
EINECS: 215-193-9
Merck Index: 12,04984

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR