Magnesium hydroxide ≥99.8%

632309-25G 632309-100G
220083-151EA 356.92 USD
220083-151 220083-150
Magnesium hydroxide ≥99.8%
Magnesium hydroxide
Formula: Mg(OH)₂
MW: 58.32 g/mol
Density: 2.3261 to 2.3631 g/cm³ (20 °C)
MDL Number: MFCD00011104
CAS Number: 1309-42-8
EINECS: 215-170-3
Merck Index: 13,05693

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR