3-Bromo-5-methoxypyridine

Warning

Synonyms: 5-Bromo-3-methoxypyridine

631817-1G 631817-5G
220083-087EA 34.46 USD
220083-087 220083-088
3-Bromo-5-methoxypyridine
3-Bromo-5-methoxypyridine
Formula: C₆H₆BrNO
MW: 188.02 g/mol
MDL Number: MFCD00234169
CAS Number: 50720-12-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR