2,6-Difluoro-4-methoxybenzaldehyde ≥97%

631809-5G 631809-1G
220083-086EA 206.08 USD
220083-086 220083-085
2,6-Difluoro-4-methoxybenzaldehyde ≥97%
2,6-Difluoro-4-methoxybenzaldehyde
Formula: C₈H₆F₂O₂
MW: 172.13 g/mol
MDL Number: MFCD04114368
CAS Number: 256417-10-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR