4-Amino-2-(trifluoromethyl)benzonitrile ≥97%

Synonyms: 4-Cyano-3-trifluoromethylaniline

631752-5G 631752-1G
220083-071EA 122.77 USD
220083-071 220083-070
4-Amino-2-(trifluoromethyl)benzonitrile ≥97%
4-Amino-2-(trifluoromethyl)benzonitrile
Formula: C₈H₅F₃N₂
MW: 186.14 g/mol
MDL Number: MFCD00042155
CAS Number: 654-70-6
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR