2-Chloro-5-iodobenzoic acid ≥97%

631736-5G 631736-25G
220083-064EA 29.08 USD
220083-064 220083-063
2-Chloro-5-iodobenzoic acid ≥97%
2-Chloro-5-iodobenzoic acid
Formula: C₇H₄ClIO₂
MW: 282.46 g/mol
MDL Number: MFCD00079731
CAS Number: 19094-56-5
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR