2-Amino-3-bromo-5-methylbenzoic acid ≥97%

631531-5G 631531-1G
220083-053EA 60.31 USD
220083-053 220083-052
2-Amino-3-bromo-5-methylbenzoic acid ≥97%
2-Amino-3-bromo-5-methylbenzoic acid
Formula: C₈H₈BrNO₂
MW: 230.06 g/mol
MDL Number: MFCD00051705
CAS Number: 13091-43-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR