(4-Bromophenyl)acetic acid ethyl ester ≥97%

Synonyms: Ethyl 2-(4-bromophenyl)acetate, Ethyl (4-bromophenyl)acetate

631353-25G 631353-5G
220083-043EA 71.08 USD
220083-043 220083-044
(4-Bromophenyl)acetic acid ethyl ester ≥97%
(4-Bromophenyl)acetic acid ethyl ester
Formula: C₁₀H₁₁BrO₂
MW: 243.1 g/mol
MDL Number: MFCD00016333
CAS Number: 14062-25-0

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR