(3,5-Dimethoxy-4-hydroxyphenyl)acetic acid ≥97%

Synonyms: 2-(4-Hydroxy-3,5-dimethoxyphenyl)acetic acid, 4-Hydroxy-3,5-dimethoxyphenylacetic acid

631310-1G 631310-5G
220083-035EA 53.85 USD
220083-035 220083-036
(3,5-Dimethoxy-4-hydroxyphenyl)acetic acid ≥97%
(3,5-Dimethoxy-4-hydroxyphenyl)acetic acid
Formula: C₁₀H₁₂O₅
MW: 212.2 g/mol
MDL Number: MFCD00016834
CAS Number: 4385-56-2
EINECS: 224-494-4

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR