3,4,5-Trimethoxyanthranilic acid ≥97%

Warning

631272-5G 631272-25G
220082-826EA 89.06 USD
220082-826 220082-825
3,4,5-Trimethoxyanthranilic acid ≥97%
3,4,5-Trimethoxyanthranilic acid
Formula: C₁₀H₁₃NO₅
MW: 227.22 g/mol
MDL Number: MFCD00051630
CAS Number: 61948-85-4
EINECS: 263-344-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR